Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3975

Ponesimod

new

≥95% (HPLC)

Synonym(s):

(2Z,5Z)-5-[3-Chloro-4-[(2R)-2,3dihydroxypropoxy ]benzylidene ]-3-(2-methylphenyl)-2-(propylimino )-1,3-thiazolidin-4-one, (R)-5-[3-Chloro-4-(2,3-dihydroxy-propoxy)-benz[Z]ylidene]-2-([Z]-propylimino)-3-o-tolyl-thiazolidin-4-one, 5-[3-Chloro-4-((2R)-2,3-dihydroxy-propoxy)-benz-(Z)-ylidene]-2-[(Z)-propylimino]-3-o-tolyl-thiazolidin-4-one, ACT 128800, ACT-128800, ACT128800

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25ClN2O4S
CAS Number:
Molecular Weight:
460.97
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

S1\C(=N/CCC)\N(C(=O)\C\1=C\c3cc(c(cc3)OC[C@H](O)CO)Cl)c2c(cccc2)C

InChI

1S/C23H25ClN2O4S/c1-3-10-25-23-26(19-7-5-4-6-15(19)2)22(29)21(31-23)12-16-8-9-20(18(24)11-16)30-14-17(28)13-27/h4-9,11-12,17,27-28H,3,10,13-14H2,1-2H3/b21-12-,25-23-/t17-/m1/s1

InChI key

LPAUOXUZGSBGDU-STDDISTJSA-N

Biochem/physiol Actions

Orally active, potent and subtype-selective sphingosine-1-phosphate (S1P) receptor S1P1 agonist.
Ponesimod (ACT-128800) is an orally active, potent and subtype-selective sphingosine-1-phosphate (S1P) receptor agonist (human S1P1/2/3/4/5 EC50 = 5.7 nM/>10 µM/105 nM/1.108 µM/59.1 nM by 35S-GTPγ binding, 6.0 nM/>10 µM/2.068 µM/1.956 µM/142 nM by 33P-S1P binding). Ponesimod oral administration causes a significant decrease in peripheral lymphocyte counts in rats in vivo (by 65-75% 3h post 3 mg/kg).

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service