SBR00076
Baricitinib
≥98% (HPLC), powder
Synonym(s):
INCB 028050, LY 3009104
About This Item
Recommended Products
Quality Level
assay
≥98% (HPLC)
form
powder
color
off-white
mode of action
enzyme | inhibits
storage temp.
2-8°C
SMILES string
CCS(=O)(=O)N1CC(C1)(CC#N)N2C=C(C=N2)C3=C4C=CNC4=NC=N3
InChI
InChI=1S/C16H17N7O2S/c1-2-26(24,25)22-9-16(10-22,4-5-17)23-8-12(7-21-23)14-13-3-6-18-15(13)20-11-19-14/h3,6-8,11H,2,4,9-10H2,1H3,(H,18,19,20)
InChI key
XUZMWHLSFXCVMG-UHFFFAOYSA-N
General description
Application
Biochem/physiol Actions
signalword
Danger
hcodes
Hazard Classifications
Repr. 1B - STOT RE 1 - STOT SE 3
target_organs
Blood,Bone marrow,lymphatic system, Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service