Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAE0218

Sigma-Aldrich

Anti-MBP Magnetic Beads

Magnetic agarose, suspension

Synonym(s):

anti Maltose binding Protein, MBP antibody, MBP epitop Tag, MBP TAG antibody, Anti-MBP antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

biological source

mouse

Quality Level

conjugate

magnetic beads

form

suspension

shelf life

2 yr (retest)

shipped in

ambient

storage temp.

2-8°C

General description

Anti-MBP Magenetic Beads are a conjugate of activated magnetic cross-linked agarose beads to a monoclonal antibody that was raised against the Maltose Binding Protein (MBP) sequence. Monoclonal Anti-Maltose Binding Protein recognizes native, as well as denatured-reduced, forms of purified MBP or MBP fusion proteins.

Application

Anti-MBP magnetic beads are useful in the purification of an MBP fusion protein in bacteria or in cells and tissues transfected with a MBP fusion protein expressing vectors. Monoclonal Anti-MBP recognizes specifically a MBP tagged fusion proteins expressed in transfected mammalian cells or produced by in vitro translation.

Binding capacity: 20-62 nmole of MBP-tagged fusion protein per 1 ml of settled resin.

Elution capacity: 5-62 nmole/mL settled resin.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service