Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501203

Sigma-Aldrich

Anti-SLC39A1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Solute carrier family 39 member 1, ZIP-1, Zinc transporter ZIP1, Zrt- and Irt-like protein 1, hZIP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 34 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC39A1(27173)

General description

Anti-SLC39A1 Antibody detects endogenous levels of total SLC39A1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human SLC39A1.

Immunogen Range: 111-160

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Segawa et al.
Life sciences, 113(1-2), 40-44 (2014-08-05)
In this study, we evaluated the effect of divalent metal cations on zinc uptake via mouse Zrt/Irt-like protein 1 (mZIP1), a ubiquitously expressed zinc transporter, which plays a role in maintenance of cellular zinc homeostasis, using HEK293T cells overexpressing it.
Xiao Dong et al.
Urologic oncology, 32(6), 885-892 (2014-06-01)
The human ZRT, IRT-like protein 1 (hZIP1) has been associated with tumorigenesis. However, its role in clear cell renal cell carcinoma (ccRCC) has not been yet reported. The objective was to investigate hZIP1 expression in ccRCC and its association with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service