MilliporeSigma
  • hZIP1 that is down-regulated in clear cell renal cell carcinoma is negatively associated with the malignant potential of the tumor.
hZIP1 that is down-regulated in clear cell renal cell carcinoma is negatively associated with the malignant potential of the tumor.
Urologic oncology (2014-06-01)
Xiao Dong, Chuize Kong, Zhe Zhang, Xiankui Liu, Bo Zhan, Zhipeng Chen, Du Shi
ABSTRACT
The human ZRT, IRT-like protein 1 (hZIP1) has been associated with tumorigenesis. However, its role in clear cell renal cell carcinoma (ccRCC) has not been yet reported. The objective was to investigate hZIP1 expression in ccRCC and its association with clinicopathological features. A total of 106 ccRCC tissue samples and corresponding normal kidney tissue samples were examined, along with 3 ccRCC cell lines (ACHN, 769-P, and 786-O). Real-time polymerase chain reaction, Western blot, and immunohistochemistry were used to investigate the expression of hZIP1 and its relationship with clinicopathological features. The ACHN cell line, exhibiting the highest hZIP1 expression, was transfected with hZIP1 small interfering RNA or mock small interfering RNA. Cellular proliferation was measured by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay. Invasion was determined by Transwell assay. The level of hZIP1 was decreased in ccRCC tissues when compared with normal tissues. hZIP1 expression significantly decreased with increasing clinical stage and pathological grade in ccRCC samples (P<0.05), showing a significant negative correlation with the histological grade (P<0.05). High hZIP1 expression was associated with a better disease-free survival (P<0.01). Silencing of hZIP1 expression enhanced the proliferative and invasive abilities of ACHN cells. Results suggest that hZIP1 may act as a tumor suppressor in ccRCC. hZIP1 is closely correlated with clinicopathological features. High hZIP1 expression may be an indicator of good prognosis in ccRCC.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%