Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500071

Sigma-Aldrich

Anti-SCARA5

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Scavenger receptor class A member 5, Anti-TESR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 24-39 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCARA5(286133)

General description

The gene SCARA5 (scavenger receptor class A member 5) is mapped to human chromosome 8p21. It belongs to the SCARA family of proteins.

Immunogen

SCARA5 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human SCARA5.

Biochem/physiol Actions

SCARA5 (scavenger receptor class A member 5) is a tumor suppressor. It plays an important role in growth regulation. It interacts with serum ferritin and helps it in crossing the blood-retinal barrier. Changes in the expression of SCARA5 are linked with retinopathy. It might also be involved in the suppression of inflammatory fibrotic responses.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500071.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luísa Mendes-Jorge et al.
PloS one, 9(9), e106974-e106974 (2014-09-27)
Iron is essential in the retina because the heme-containing enzyme guanylate cyclase modulates phototransduction in rods and cones. Transferrin endocytosis is the classical pathway for obtaining iron from the blood circulation in the retina. However, the iron storage protein ferritin
Richard D Kenagy et al.
Journal of vascular surgery, 64(1), 202-209 (2015-05-04)
Approximately 30% of autogenous vein grafts develop luminal narrowing and fail because of intimal hyperplasia or negative remodeling. We previously found that vein graft cells from patients who later develop stenosis proliferate more in vitro in response to growth factors than
Y V Sanders et al.
Journal of thrombosis and haemostasis : JTH, 13(6), 956-966 (2015-04-03)
von Willebrand factor (VWF) levels in healthy individuals are influenced by variations in genetic loci other than the VWF gene, whose contribution to VWF levels in patients with von Willebrand disease (VWD) is largely unknown. To investigate the association between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service