MilliporeSigma
  • CLEC4M and STXBP5 gene variations contribute to von Willebrand factor level variation in von Willebrand disease.
CLEC4M and STXBP5 gene variations contribute to von Willebrand factor level variation in von Willebrand disease.
Journal of thrombosis and haemostasis : JTH (2015-04-03)
Y V Sanders, J G van der Bom, A Isaacs, M H Cnossen, M P M de Maat, B A P Laros-van Gorkom, K Fijnvandraat, K Meijer, C M van Duijn, E P Mauser-Bunschoten, J Eikenboom, F W G Leebeek
ABSTRACT
von Willebrand factor (VWF) levels in healthy individuals are influenced by variations in genetic loci other than the VWF gene, whose contribution to VWF levels in patients with von Willebrand disease (VWD) is largely unknown. To investigate the association between single-nucleotide polymorphisms (SNPs), VWF levels, and bleeding phenotype. In 364 type 1 VWD and 240 type 2 VWD patients from the nationwide cross-sectional 'Willebrand in The Netherlands' (WiN) study, we studied the association between eight SNPs in STXBP5, SCARA5, ABO, VWF, STAB2, STX2, TC2N, and CLEC4M, and VWF antigen (VWF:Ag), VWF activity (VWF:Act), and bleeding phenotype as assessed with the Tosetto bleeding score. In type 1 patients, STXBP5 was associated with a lower VWF:Ag level (adjusted difference of -3.0 IU dL(-1) per allele; 95% confidence interval [CI] -6.0 to 0.1) and CLEC4M with both a lower VWF:Ag level (-4.3 IU dL(-1) per allele; 95% CI -7.9 to -0.6) and lower VWF:Act (-5.7 IU dL(-1) per allele; 95% CI -10.9 to -0.5). In type 2 patients, none of the SNPs was associated with VWF levels. None of the genetic variants was associated with bleeding score. Genetic variations in STXBP5 and CLEC4M are associated with VWF level variation in type 1 VWD, but not in type 2 VWD. This study increases our understanding of the pathophysiology of VWD, and provides a further indication of the involvement of STXBP5 and CLEC4M in determining VWF levels in VWD.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O