Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1409845

Sigma-Aldrich

Monoclonal Anti-CAMK2B antibody produced in mouse

clone 6D6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAM2, CAMK2, CAMKB, MGC29528

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D6, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.52 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2B(816)

General description

The product of this gene belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a beta chain. It is possible that distinct isoforms of this chain have different cellular localizations and interact differently with calmodulin. Eight transcript variants encoding eight distinct isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

CAMK2B (NP_742078, 405 a.a. ~ 502 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
FEPEALGNLVEGMDFHRFYFENLLAKNSKPIHTTILNPHVHVIGEDAACIAYIRLTQYIDGQGRPRTSQSEETRVWHRRDGKWQNVHFHCSGAPVAPL

Biochem/physiol Actions

Calcium/calmodulin dependent protein kinase II β (CAMK2B) may bind to calmodulin. It has been associated with major depressive disorder.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Possible inhibitor from traditional Chinese medicine for the ? form of calcium-dependent protein kinase type II in the treatment of major depressive disorder.
Hung TC
BioMed Research International, 761849-761849 (2014)
Molecular cloning and sequence analyses of calcium/calmodulin-dependent protein kinase II from fetal and adult human brain. Sequence analyses of human brain calciuum/calmodulin-dependent protein kinase II.
Li G
Molecular Biology Reports, 28(1), 35-41 (2001)
Fine-mapping of vitiligo susceptibility loci on chromosomes 7 and 9 and interactions with NLRP1 (NALP1).
Jin Y
The Journal of Investigative Dermatology, 130(3), 774-783 (2010)
Ju Hee Kim et al.
BMB reports, 44(8), 523-528 (2011-08-30)
To identify novel genes that are regulated by promoter methylation, a combinational approach involving in silico mining followed by molecular assay was performed. From the expression microarray data registered in the European bioinformatics institute (EBI), genes showing downregulation in breast
Tamara Berdyyeva et al.
PloS one, 9(11), e112068-e112068 (2014-11-06)
Therapeutic drugs for cognitive and psychiatric disorders are often characterized by their molecular mechanism of action. Here we demonstrate a new approach to elucidate drug action on large-scale neuronal activity by tracking somatic calcium dynamics in hundreds of CA1 hippocampal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service