Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADCMXRJ10TLA

Mobius® ADC Reactor 10L Carrier ASME

new

Stainless Steel Jacketed Carrier with Temperature Sensor and Load Cells, ASME certified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

stainless steel hardware

Quality Level

description

Storage temperature: 0-40°C

product line

Mobius®

parameter

4-60 °C temp. range (39-140 °F)

capacity

10 L

application(s)

pharma/biopharma processes

General description

The Mobius® ADC Reactor is a single-use mixing system, specifically designed for the key conjugation step to manufacture Antibody Drug Conjugates (ADCs).

This dedicated solution brings all the benefits from using single-use to the conjugation step. These include achieving faster turnaround times, avoiding cross-contamination risks, cleaning procedures and validation, while maintaining high product quality with no compromise on process continuity and operator safety.

Application

Specifically designed for the key conjugation step to manufacture Antibody Drug Conjugates (ADCs).

Packaging

1 carrier per package.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service