Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADCCBC1V1S

Ready To Connect Control Box 10L ADC

new

115/230V with pH & Conductivity Sensors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

material

stainless steel hardware

Quality Level

description

Storage temperature: 0-40°C

product line

Mobius®

parameter

4-60 °C temp. range (39-140 °F)

application(s)

pharma/biopharma processes

Looking for similar products? Visit Product Comparison Guide

General description

The Mobius® ADC Reactor can be monitored and controlled locally or remotely.
Plug the ready to connect control box to the network and use Ethernet IP communication protocol to monitor speed, weight, temperature, pH and conductivity remotely.

The ready to connect control box is a separate box on a four wheels chassis that can be easily moved around the Mobius® ADC Reactor carrier.

Application

Specifically designed for the key conjugation step to manufacture Antibody Drug Conjugates (ADCs).

Packaging

1 control box per package.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service