GF54633630
Nickel
tube, 1000mm, outside diameter 1.5mm, inside diameter 1.4mm, wall thickness 0.05mm, hard, 99.5%
Synonym(s):
Nickel, NI007252
About This Item
assay
99.5%
form
tubes
manufacturer/tradename
Goodfellow 546-336-30
resistivity
6.97 μΩ-cm, 20°C
L × wall thickness
1000 mm × 0.05 mm
O.D. × I.D.
1.5 mm × 1.4 mm
bp
2732 °C (lit.)
mp
1453 °C (lit.)
density
8.9 g/mL at 25 °C (lit.)
SMILES string
[Ni]
InChI
1S/Ni
InChI key
PXHVJJICTQNCMI-UHFFFAOYSA-N
General description
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Skin Sens. 1 - STOT RE 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service