Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407729

Sigma-Aldrich

Anti-PCDHA6 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~102.7 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDHA6(56142)

General description

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. (provided by RefSeq)

Immunogen

PCDHA6 (NP_061732.1, 1 a.a. ~ 950 a.a) full-length human protein.

Sequence
MVFTPEDRLGKQCLLLPLLLLAAWKVGSGQLHYSVPEEAKHGTFVGRIAQDLGLELAELVPRLFRMASKDREDLLEVNLQNGILFVNSRIDREELCGRSAECSIHLEVIVDRPLQVFHVDVEVRDINDNPPLFPVEEQRVLIYESRLPDSVFPLEGASDADVGSNSILTYKLSSSEYFGLDVKINSDDNKQIGLLLKKSLDREEAPAHNLFLTATDGGKPELTGTVQLLVTVLDVNDNAPTFEQSEYEVRIFENADNGTTVIRLNASDRDEGANGAISYSFNSLVAAMVIDHFSIDRNTGEIVIRGNLDFEQENLYKILIDATDKGHPPMAGHCTVLVRILDKNDNVPEIALTSLSLPVREDAQFGTVIALISVNDLDSGANGQVNCSLTPHVPFKLVSTFKNYYSLVLDSALDRESVSAYELVVTARDGGSPSLWATASLSVEVADMNDNAPAFAQPEYTVFVKENNPPGCHIFTVSARDADAQENALVSYSLVERRVGERALSSYISVHAESGKVYALQPLDHEELELLQFQVSARDAGVPPLGSNVTLQVFVLDENDNAPALLAPRVGGTGGAVSELVPRSLGAGQVVAKVRAVDADSGYNAWLSYELQPPASSARFPFRVGLYTGEISTTRVLDEADSPRHRLLVLVKDHGEPALTATATVLVSLVESGQAPKASSRASVGAAGPEAALVDVNVYLIIAICAVSSLLVLTLLLYTALRCSAPPTEGACTADKPTLVCSSAVGSWSYSQQRRQRVCSGEGPPKMDLMAFSPSLSPCPIMMGKAENQDLNEDHDAKPRQPNPDWRYSASLRAGMHSSVHLEEAGILRAGPGGPDQQWPTVSSATPEPEAGEVSPPVGAGVNSNSWTFKYGPGNPKQSGPGELPDKFIIPGSPAIISIRQEPTNSQIDKSDFITFGKKEETKKKKKKKKGNKTQEKKEKGNSTTDNSDQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service