Skip to Content
Merck
All Photos(1)

Key Documents

EMU012821

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Sphk1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCAGAAACCCCTGTGTAGCCTCCCTGGAGGCTCCGGCAATGCGCTGGCAGCTTCTGTGAACCACTATGCTGGGTACGAGCAGGTGACTAATGAAGACCTGCTCATCAACTGCACACTGCTGTTGTGCCGCCGGCGCCTGTCACCCATGAACCTGCTGTCCCTGCACACTGCTTCTGGGCTGCGGCTCTATTCTGTGCTCAGTCTGTCCTGGGGCTTTGTTGCTGACGTGGACCTCGAGAGTGAGAAGTACAGGCGCTTGGGGGAGATTCGTTTCACAGTGGGCACCTTCTTTCGCCTAGCAAGCCTGCGCATCTACCAAGGCCAACTGGCCTACCTTCCTGTAGGAACTGTGGCCTCTAAGAGACCCGCCTCTACACTGGTGCAGAAGGGCCCCGTCGACACACACCTTGTTCCTCTGGAGGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujin Zhang et al.
The Journal of clinical investigation, 124(6), 2750-2761 (2014-05-20)
Sphingosine-1-phosphate (S1P) is a bioactive lipid that regulates multicellular functions through interactions with its receptors on cell surfaces. S1P is enriched and stored in erythrocytes; however, it is not clear whether alterations in S1P are involved in the prevalent and
Se Kyoo Jeong et al.
Journal of dermatological science, 79(3), 229-234 (2015-06-27)
The ceramide metabolite, sphingosine-1-phosphate (S1P), regulates multiple cellular functions in keratinocytes (KC). We recently discovered that production of a key innate immune element, cathelicidin antimicrobial peptide (CAMP), is stimulated via a NF-κB-dependent mechanism that is activated by S1P when S1P

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service