Skip to Content
Merck
All Photos(2)

Key Documents

V800123

Sigma-Aldrich

Cobalt(II) acetate

LR, 31.3-35.3%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)2Co
CAS Number:
Molecular Weight:
177.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
solid

grade

LR

product line

Vetec

form

solid

concentration

31.3-35.3%

mp

298 °C (dec.) (lit.)

SMILES string

CC([O-])=O.[Co+2]

InChI

1S/2C2H4O2.Co/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

QAHREYKOYSIQPH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihisa Kimura et al.
The European journal of neuroscience, 41(10), 1276-1293 (2015-03-27)
The thalamic reticular nucleus (TRN) occupies a highly strategic position to modulate sensory processing in the thalamocortical loop circuitries. It has been shown that TRN visual cells projecting to first- and higher-order thalamic nuclei have distinct levels of burst spiking
Owen M Williams et al.
Dalton transactions (Cambridge, England : 2003), 44(29), 13017-13029 (2015-04-14)
Reported here are the syntheses and characterization of cobaloximes that feature a bis(imino)acenaphthene (BIAN) appended ligand. The X-ray crystal structures and spectroscopy ((1)H NMR or EPR) of the complexes within the series [Co(aqdBF2)2(MeCN)2] (), [Co(aqdBF2)(MeCN)2](-) () and [Co(aqdBF2)2(MeCN)2](2-) (, )
Mohamed E Mahmoud et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 50(10), 1072-1081 (2015-06-30)
In this work, dioctyl phthalate (Dop) was used as a highly plasticizing material to coat and link the surface of basic cellulose (Cel) with baker's yeast for the formation of a novel modified cellulose biosorbent (Cel-Dop-Yst). Characterization was accomplished by
Chih-Yu Hsu et al.
ACS applied materials & interfaces, 7(33), 18266-18272 (2015-08-01)
Black-to-transmissive electrochromism has been obtained with a Co(II)-based metallo-supramolecular polymer (polyCo). Thin films of polyCo, based on bisterpyridine ligand assembled with Co(II) metal ion, were constructed by spray casting the polymer onto ITO glass. With such simple fabricating means to
Jung-Che Tsai et al.
Chemistry, an Asian journal, 10(9), 1932-1939 (2015-07-15)
Mesoporous cobalt sulfide nanotube arrays on FTO-coated glass were synthesized by combining three simple technologies: the selective etching of ZnO sacrificial templates, mesoporous Co3 O4 formation from cobalt-chelated chitosan, and ion-exchange reaction (IER). The mesoporous Co3 O4 nanotubes composed of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service