Merck
  • A novel cellulose-dioctyl phthate-baker's yeast biosorbent for removal of Co(II), Cu(II), Cd(II), Hg(II) and Pb(II).
A novel cellulose-dioctyl phthate-baker's yeast biosorbent for removal of Co(II), Cu(II), Cd(II), Hg(II) and Pb(II).
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2015-06-30)
Mohamed E Mahmoud, Amr A Yakout, Marwa T Abed El Aziz, Maher M Osman, Tarek M Abdel-Fattah
ABSTRACT
In this work, dioctyl phthalate (Dop) was used as a highly plasticizing material to coat and link the surface of basic cellulose (Cel) with baker's yeast for the formation of a novel modified cellulose biosorbent (Cel-Dop-Yst). Characterization was accomplished by Fourier Transform Infrared Spectroscopy (FT-IR), Thermogravimetric analysis (TGA) and Scanning Electron Microscope (SEM) measurements. The feasibility of using Cel-Dop-Yst biosorbent as an efficient material for removal of Co(II), Cu(II), Cd(II), Hg(II) and Pb(II) ions was explored using the batch equilibrium technique along with various experimental controlling parameters. The optimum pH values for removal of these metal ions were characterized in the range of 5.0-7.0. Cel-Dop-Yst was identified as a highly selective biosorbent for removal of the selected divalent metal ions. The Cel-Dop-Yst biosorbent was successfully implemented in treatment and removal of these divalent metal ions from industrial wastewater, sea water and drinking water samples using a multistage microcolumn technique.
Sigma-Aldrich
Copper(II) acetate, LR, ≥98%
Sigma-Aldrich
Cobalt(II) acetate, LR, 31.3-35.3%
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) acetate, 98%
Sigma-Aldrich
Cobalt(II) acetate, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Potassium bromide, LR, ≥99%
Sigma-Aldrich
Potassium chloride, LR, ≥99%
Sigma-Aldrich
Potassium bromide, AR, ≥99.5%
Sigma-Aldrich
Potassium chloride, AR, ≥99.5%
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Potassium chloride, BioUltra, ≥99.5% (AT)