1692710
USP
Trimipramine Related Compound A
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
5-[3-(Dimethylamino)-2-methylpropyl]-5H-dibenz[b,f]azepine, N,Nβ-Trimethyl-5H-dibenz[b,f]azepine-5-propanamine
About This Item
grade
pharmaceutical primary standard
API family
trimipramine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
N1(c2c(cccc2)C=Cc3c1cccc3)CC(CN(C)C)C
InChI
1S/C20H24N2/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22/h4-13,16H,14-15H2,1-3H3
InChI key
RNFADUFBCJRRPS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service