Skip to Content
Merck
Trimipramine Related Compound A pharmaceutical secondary standard, certified reference material

Trimipramine Related Compound A

Trimipramine Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
5-[3-(Dimethylamino)-2-methylpropyl]-5H-dibenz[b,f]azepine, N,Nβ-Trimethyl-5H-dibenz[b,f]azepine-5-propanamine
Empirical Formula (Hill Notation):
C20H24N2
CAS Number:
Molecular Weight:
292.42

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade