Skip to Content
Merck
All Photos(1)

Key Documents

74924

Supelco

5-Amino-4-chloro-3-pyridazinone

PESTANAL®, analytical standard

Synonym(s):

Chloridazon-desphenyl Metabolite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4ClN3O
CAS Number:
Molecular Weight:
145.55
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0%

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

Clc1c(nncc1N)O

InChI key

FEWPCPCEGBPTAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D, and Analytical use only. Not for drug, household or other uses.
Metabolite for water testing

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service