Skip to Content
Merck
5-Amino-4-chloro-3-pyridazinone PESTANAL®, analytical standard

5-Amino-4-chloro-3-pyridazinone

5-Amino-4-chloro-3-pyridazinone PESTANAL®, analytical standard
Synonym(s):
Chloridazon-desphenyl Metabolite
Empirical Formula (Hill Notation):
C4H4ClN3O
CAS Number:
Molecular Weight:
145.55

Products

Brand
Formula Weight
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing