Skip to Content
Merck
All Photos(1)

Key Documents

50114-U

Supelco

Purospher® STAR RP-18 Endcapped (2 µm) Capillary Column

L × I.D. 50 mm × 0.3 mm, pk of 1 ea

Synonym(s):

Purospher® STAR RP-18 Endcapped (2 µm) Capillary Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

Quality Level

Agency

suitable for USP L1

description

UHPLC Column

feature

endcapped

packaging

pk of 1 ea

extent of labeling

17% C loading

parameter

10000 psi max. pressure (700 bar)
65 °C temp. limit

technique(s)

LC/MS: suitable
UHPLC: suitable

L × I.D.

50 mm × 0.3 mm

matrix

fully porous particle support

matrix active group

C18 bonding phase

particle size

2 μm

pore size

120 Å pore size

operating pH

1.5-10.5

separation technique

reversed phase

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, catecholamines, explosives, oligonucleotides, esters, fat soluble vitamins, lipids, fatty acids, flavonoids, ketones, nitrosamines, nucleosides, PAHs, PCBs, peptides, pesticides, phenols, phthalates, preservatives, steroids, metabolized steroids, sulfonamides, sweeteners, and water soluble vitamins.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Outstanding pH stability from pH 1.5 to 10.5
  • No phase collapse when using highly aqueous mobile phases
  • Good suitability for LC-MS applications

Legal Information

Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service