Skip to Content
Merck
All Photos(1)

Key Documents

SCM064SFM

LX-2 Human Hepatic Stellate Serum-Free Media

new

Synonym(s):

HSC, Hepatic lipocytes, Hepatic pericytes, Ito cells, Perisinusoidal cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human

Quality Level

packaging

vial of 250 mL

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Application

Appearance (Color): Clear, Red Liquid
pH: 7.2 – 7.8
Osmolality: 289-310 mOsm
Sterility Tested: No Growth/Pass
Endotoxin: 2 EU/mL
Mycoplasma: Negative
Functional Assay: Thaw and culture of LX-2 cells.

Features and Benefits

Serum-Free media optimized to expand and culture LX-2 human hepatic stellate cells.

Target description

LX-2 Human Hepatic Stellate Serum-Free Media (SCM064SFM) is a ready to use, serum-free culture medium specifically designed for the growth and proliferation of LX-2 human hepatic stellate cells (SCC064, SCC064SF).

Hepatic stellate cells are a major cell type responsible for liver fibrosis following their activation into fibrogenic myofibroblast-like cells in diseases such as chronic alcoholism, hepatitis B and C, fatty liver disease, obesity, and diabetes. LX-2 cells, an immortalized human hepatic stellate cells, retain key features of primary hepatic stellate cells (HSCs) and have been extensively utilized in the study of liver fibrosis.

SCM064SFM eliminates the reliance on FBS, providing a consistent and controlled environment for cell culture. This ensures reproducibility in experimental results and reduces variability often associated with serum-based media. The media is optimized to support the growth and proliferation of LX-2 cells, enabling researchers to conduct experiments with similar cell yields and viability compared with cells grown on FBS supplemented media. By eliminating the need for FBS, SCM064SFM significantly reduces the demand for animal-derived products, contributing to ethical research practices and animal welfare.

Storage and Stability

Upon receipt store at -20˚C. When ready to use, thaw overnight at 2-8˚C. Once thawed, mix thoroughly. Use immediately and store at 2-8˚C for up to 3-4 weeks. Multiple freeze/thaw cycles are not recommended.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service