Skip to Content
Merck
All Photos(1)

Key Documents

SCC064SF

LX-2 Hepatic Stellate Cell Line-SF Adapted

new

Synonym(s):

HSC, Hepatic lipocytes, Hepatic pericytes, Ito cells, Perisinusoidal cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human

Quality Level

packaging

vial of ≥1 x 10^6 vial (viable cells per vial)

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases tested by using the Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of human origin and negative for interspecies contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Features and Benefits

Serum-Free media adapted LX-2 human hepatic stellate cells that been extensively characterized and retain key features of hepatic stellate cytokine signaling, neuronal gene expression, retinoid metabolism, and fibrogenesis , making them a highly suitable model of human hepatic fibrosis.

Target description

Hepatic stellate cells are a major cell type responsible for liver fibrosis following their activation into fibrogenic myofibroblast-like cells in diseases such as chronic alcoholism, hepatitis B and C, fatty liver disease, obesity and diabetes. There is an increasing need for renewable cell culture models that faithfully recapitulate their in vivo phenotype, particularly for human studies.

LX-2 was generated by immmortalization of primary human hepatic stellate cells with the SV40 large T antigen followed by selective culture of early passaged cells in low serum media conditions.

Immortalized LX-2 was established by Xu et al to overcome issues of culture variability and to provide a stable and unlimited source of human hepatic stellate cells that are homogeneous. These cell lines have been extensively characterized and retain key features of cytokine signaling, neuronal gene expression, retinoid metabolism, and fibrogenesis, making them highly suitable for culture based studies of human hepatic fibrosis.

The LX-2 Hepatic Stellate Cell Line-SF Adapted cell ine (SCC064SF) has been preadapted to serum-free media conditions using the LX-2 Human Hepatic Stellate Serum-Free Media (SCM064SFM). It is recommended to use SCM064SFM to thaw and expand the cell line.

Storage and Stability

LX-2 cells should be stored in liquid nitrogen. The cells can be passage for at least 10 passages without significantly affecting the cell marker expression and functionality. LX-2 cells have been successfully expanded past passage 50 in culture.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service