Skip to Content
Merck
All Photos(1)

Key Documents

MSTFA102A

Millipore

MAST® Sanitant Pre-Filter Assembly

new

For use with MAST® Autosampling Solution

Synonym(s):

MAST Filter, Pre-Filter Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

storage condition

room temperature

General description

The MAST® Sanitant Pre-Filter Assembly is designed for compatibility with the MAST® Autosampling Solution through its connection with the Cleaning and Sanitation station. The pre-filter assembly shields the system from any particles that may exist in the sanitant, amplifying the protection provided by the MAST® Sanitant Filter Assembly and MAST® Purge Filter ASsembly.

Application

MAST® Autosampling Solution
Automated Sampling, Process Analytical Technologies (PAT), Bioprocessing

Features and Benefits

  • Reduced contamination risk
  • User friendly
  • Single Use

Packaging

12 units per box

Other Notes

To be replaced monthly

Legal Information

MAST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service