MSTFA101A
MAST® Purge Filter Assembly

For use with MAST® Autosampling Solution
Synonym(s):
MAST Filter, PurgeFilter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
Quality Level
storage condition
room temperature
General description
The MAST® Purge Filter Assembly is attached to the Sample Pilot, safeguarding both the Sample Pilot and the sample source against contamination.
Application
MAST® Autosampling Solution
Automated Sampling, Process Analytical Technologies (PAT), Bioprocessing
Features and Benefits
- Reduced contamination risk
- User friendly
- Single Use
Packaging
10 units per box
Preparation Note
Autoclaved by User following connection to Sample Pilot
Other Notes
To be replaced after every autoclave cycle
Legal Information
MAST is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service