DWK293011908
Tamper-Proof Screw Caps For Narrow Neck Square Bottles
neck Joints: (GL 32 H)
Synonym(s):
293011908
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
Recommended Products
material
blue polypropylene screw cap
Joint
Joints: neck (GL 32 H)
packaging
pkg of 10 ea
manufacturer/tradename
293011908
Looking for similar products? Visit Product Comparison Guide
Application
Sterile liquid transfer between containers, Liquid sampling into bottles from larger containers, Port assemblies for spinner flasks, and cell culture vessels,
Features and Benefits
for lime soda bottle GL 32
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service