Skip to Content
Merck
All Photos(2)

Key Documents

DWK293021909

Safety Tamper-Proof Screw Caps For Narrow Neck Square Bottles

neck Joints: (GL 32 H)

Synonym(s):

293021909

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

material

red polypropylene screw cap

Joint

Joints: neck (GL 32 H)

packaging

pkg of 10 ea

manufacturer/tradename

293021909

Looking for similar products? Visit Product Comparison Guide

Application

Sterile liquid transfer between containers, Liquid sampling into bottles from larger containers, Port assemblies for spinner flasks, and cell culture vessels, safety screw

Features and Benefits

for lime soda bottle GL 32

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service