주요 문서
57626
Sample Loop for Model 7725 Injector
volume 50 μL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
용적
50 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Sample loop for model 7725 injector (volume 50 μL) is suitable for advanced fluid-handling solutions for a wide variety of analytical instrumentation and in vitro diagnostic systems. It can be utilized in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)