주요 문서
57417
Supelco
Gripper fitting
for use with 1/16 in. tubing
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12352200
설명
Tube end fittings (nuts) not included.
포장
pkg of 10 ea
기술
LPLC: suitable
호환성
for use with 1/16" tubing
for use with Omnifit fittings
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Gripper fitting is used in sealing of the protein chip cartridge by screwing the plastic holder and connection tubing.
애플리케이션
Gripper fitting may be used in chromatographic sample preparation, purification and traditional/classical flash chromatography.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Kevin A Heyries et al.
Biosensors & bioelectronics, 23(12), 1812-1818 (2008-04-09)
Protein microarrays for allergen-specific antibodies detection were integrated in microfluidic chips, with imaging chemiluminescence as the analytical technique. This paper demonstrates the feasibility of miniaturized chemiluminescent ELISA by presenting rapid, reproducible and sensitive detection of protein antibodies using microfluidics. Three