주요 문서
29112-U
Supelco
Inter-Seal bonded caps (inseparable caps and liner)
black cap, PTFE/silicone septa, thread for 10-425
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
NACRES:
NB.11
material
PTFE/silicone septa
black cap
포장
pkg of 100 ea
피팅
thread for 10-425
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Inter-Seal bonded caps eliminate expensive and time consuming hand assembly while preventing liner fallout. They are made by a bonding process that bonds PTFE/silicone into polypropylene closures without using adhesives. The bonding properly seats and secures the septa within the vial opening. This integrated one piece system withstands multiple injections, making it very cost effective.
Benefits include:
  • Significant bond strength
  • Withstands multiple injections
  • Resistant to coring
  • Excellent resealability
  • Broad chemical resistance
법적 정보
Inter-Seal is a trademark of Integrated Liner Technologies, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)