주요 문서
Z370959
Sigma-Aldrich
Dialysis tubing closures
white, size 50 mm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41123102
NACRES:
NB.22
material
white
크기
50 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Clamping closures gently lock over dialysis tubing, providing a leakproof seal without knots. Can be used with cellulose ester, regenerated cellulose, and PVDF dialysis membranes. Made entirely of Nylon 66, closures have an operating temperature range of 0-90°C, and can be sterilized with ethylene oxide, γ-irradiation, ethanol, or formalin.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)