주요 문서
Z326615
BRAND® BLAUBRAND® volumetric flask, PP stopper, clear glass
volume 200 mL, accuracy: 0.15 mL, polypropylene stopper, neck joint: ST/NS 14/23
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
material
borosilicate glass
polypropylene stopper
조인트
joint: ST/NS neck14/23
설명
calibrated to contain (TC, IN)
무균
non-sterile
특징
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX))
closure type PP Stopper
포장
pack of 1
제조업체/상표
BRAND
부피 측정 범위
200 mL, accuracy: 0.15 mL
용량
200 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3. Supplied with a square-grip PP stopper. Marks and inscriptions in high contast blue enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
기타 정보
BR37250 (India)
법적 정보
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
프로토콜
This page contains detailed information on working with volumetric flasks and how to prepare a standard solution with a volumetric flask.