주요 문서
TMO167525
Nunc® Cell Factory(TM) filter adaptor cap
case of 100 ea
동의어(들):
cell harvesting system, closed cell culture system
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41161502
NACRES:
NB.24
포장
case of 100 ea
제조업체/상표
Nunc 167525
일반 설명
Further the development of applications that require a more closed cell culture system using Thermo Scientific Nunc Cell Factory Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
법적 정보
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)