주요 문서
R8133
Rulers for autoradiography
can be used with preflashed film and intensifying screens
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41105341
NACRES:
NB.31
제조업체/상표
Diversified Biotech ARS-150
일반 설명
Non-radioactive phosphorescent emission intensity is adjusted to provide sharp markings on autoradiograms and subsequent photographs. Visible under normal lighting. Supplied with adhesive backing. Can be used with preflashed film and intensifying screens.
포장
Pkg. contains four 11 × 200 mm rulers.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)