주요 문서
GE17-0572-02
Sephadex® G-25 DNA GR
Cytiva 17-0572-02, pack of 100 g
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
23151817
NACRES:
NA.52
포장
pack of 100 g
제조업체/상표
Cytiva 17-0572-02
저장 조건
(.01 M NaOH)
입자 크기
20-80 μm (dry)
작동 범위
2-13
관련 카테고리
일반 설명
illustra Sephadex® G-25 DNA grade SF.
특징 및 장점
  • For purification of DNA ( 10 bases in length) from small molecules by gel filtration.
  • Tested to ensure reproducibly high recovery of DNA.
  • Used in MicroSpin G-25 Columns and NAP Columns.
  • Enables self preparation of columns for nucleic acid purification.
저장 및 안정성
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
분석 메모
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
법적 정보
MicroSpin is a trademark of Cytiva
NAP is a trademark of Cytiva
Sephadex is a registered trademark of Cytiva
illustra is a trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서
This page shows how to solve practical problems that may occur when running an affinity chromatography column.
프로토콜
This page shows describes fractional precipitation for removing gross impurities from affinity chromatography samples with small volumes.
This page shows how to perform a separation with a Sephadex column from GE Healthcare which are Size Exclusion Chromatography media consisting of an epichlorohydrin cross-linked dextran.
This page shows how to prepare samples for purification with affinity chromatography.
관련 콘텐츠
Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.