주요 문서
BAB602150000
H-B® Durac® Thermometer
temperature -20 - 150 °C (0-300 F), Maximum Registering, calibration
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41112213
NACRES:
NB.79
설명
usage autoclave thermometer
특징
calibration
Maximum Registering
포장
pack of 1 ea
제조업체/상표
Bel-Art B60215-0000
온도 측정 범위
-20 - 150 °C (0-300 F)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Durable bi-metal thermometer has a maximum registering pointer (red) that is moved up by the temperature measuring pointer (black) to indicate the highest temperature achieved, and remains there until manually reset.
특징 및 장점
  • Threaded connection (1/4” NPT) screws directly into in-line processes; can be used to mount thermometer on included stand
  • Waterproof and pressurized; withstands autoclave cycles and has a removable protective silicone sleeve to prevent moisture from making contact with the dial
  • Constructed from durable 304 stainless steel with a glass lens; withstands pressure, high temperatures and rust
  • Probe length: 50mm (2")
법적 정보
Durac is a registered trademark of SP Scienceware, a division of SP
H-B is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)