주요 문서
1.12938
Supelco
Tap (PE)
self-venting, for 3/4 in. vessels internal thread 2" to 3/4" reducer supplied
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122409
NACRES:
NB.51
피팅
for 3/4 in. vessels internal thread 2" to 3/4" reducer supplied
저장 온도
15-25°C
일반 설명
self-venting, for vessels with 3/4" internal thread (2" to 3/4" reducer supplied)
Storage Class Code