주요 문서
BAH327870000
SP Bel-Art® Carboy Pump
for use with 20 L (5 Gal) carboy
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.35
포장
pack of 1 ea
제조업체/상표
Bel-Art H32787-0000
관련 카테고리
일반 설명
The Poly-Hand Pump threads directly to a standard drum bung or carboy. It is constructed entirely of rugged polyethylene and is inert to strong acids, alkalis and many solvents.
특징 및 장점
  • Delivers by positive pressure on the downstroke of the piston; does not pressurize the vessel
  • Ergonomic ‘T’ handle fits comfortably in a gloved hand
  • Pump will efficiently deliver against a 3.65m (12ft) head of water and empty a drum or carboy to within 3 mm (⅛”) of the bottom
  • Sizes are available to fit 114L and 220L (30/50gal) plastic or steel drums and 20L (5gal) carboy (F10795-0000)
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)