주요 문서
Z743164
Julabo® bath covers
flat cover to fit CORIO C-B19/27, CD-B19/27 and CD-BT19/27
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.77
material
stainless steel
제조업체/상표
Julabo 9970291
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Stainless Steel Flat bath cover for 19L and 27L bath tanks. The use of a bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath. Suitable for use with Julabo® CORIO C-B19, C-B27, CD-B19, CD-B27, CD-BT19, CD-BT27.
특징 및 장점
  • reduces evaporation
  • prevents contamination of samples
법적 정보
CORIO is a trademark of Julabo GmbH
Julabo is a registered trademark of Julabo USA, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)