주요 문서
Z743146
Julabo® shaking water baths, installation cooling coil, for SW 22/23
pack of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103706
NACRES:
NB.77
포장
pack of 1 ea
제조업체/상표
Julabo 8970415
일반 설명
Installation cooling coil (5mm outer diameter) can be used for temperature control tasks close to ambien temperature. A hose with an inner diameter of 6/8 mm is required for this. The cooling coil is screwed into the water bath through the drain cock.
법적 정보
Julabo is a registered trademark of Julabo USA, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)