주요 문서
Z684546
Ace Rotary Evaporator Vial Adapter Kit
FETFE® O-ring, adapter joint: ST/NS 24/40
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.35
material
FETFE® O-ring
조인트
joint: ST/NS adapter24/40
설명
PTFE adapters with #15 Ace-Thred to GPI vial thread, sizes 8-425, 13-425, 15-425, 18-400, 20-400, 22-400, 24-410
제조업체/상표
Ace Glass 13290-55
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
PTFE connecting adapters and glass conversion adapters for connecting GPI threaded vials to a rotary evaporator vapor tube or trap. Both are suitable for vacuum work. PTFE connecting adapters are available with FETFE® or Chemraz® O-rings. Order Chemraz if you are using methylene chloride or acetone.
법적 정보
Ace-Thred is a trademark of Ace Glass, Inc.
Chemraz is a registered trademark of Greene, Tweed of Delaware, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)