주요 문서
Z567256
Ace round-bottom pressure flask with thermowell
with Ace-Thred 15 PTFE front-seal plug, capacity 50 mL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
single-neck
무균
non-sterile
특징
closure type PTFE plug seal
포장
pack of 1
제조업체/상표
Ace Glass 8417-03
용량
50 mL
직경
50 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Heavy-wall borosilicate glass flasks are pressure rated to 60 psig at 120°C. The rounded bottom facilitates use in heating mantles and for other round-bottom flask applications. Flasks are supplied complete with #15 Ace-Thred PTFE front seal bushing and FETFE® O-ring.
Replacement FETFE O-rings:
Ace-Thred #Front SealBack Seal
7 Z504661 Z504742
15 Z504858 Z505021
25 Z505056 Z505110
CAUTION: This glassware is not guaranteed against breakage caused by pressure or vacuum. These flasks should never be used if scratched or otherwise damaged.
법적 정보
FETFE is a registered trademark of Ace Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)