주요 문서
Z417394
Sigma-Aldrich
Aldrich® System 45 three-hole delivery cap
PTFE, thread for 45 mm, pkg of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.43
material
PTFE
포장
pkg of 1 ea
피팅
thread for 45 mm
일반 설명
Cap has 3 holes with 1/4-28 threads at the top for use with 1/8 in. O.D. tubing. No need to precut tubing to reach the reservoir bottom, just a quick quarter turn of the lock nut (from outside the reservoir) allows positioning of PTFE tubing at any depth. Thread size: 45 mm.
법적 정보
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)