주요 문서
Z136107
Sigma-Aldrich
Scienceware® pipette support rack
holds 50 pipettes vertically in a 5 × 10 arrangement
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122801
NACRES:
NB.01
material
polypropylene
특징
Holds 50x pipettes
포장
pack of 1 ea
제조업체/상표
Bel-Art F18953-0000
길이 × 너비 × 높이
8.375 cm × 4.5 cm × 8.75 cm
일반 설명
Molded PP rack is autoclavable (121°C). Holds 50 pipettes vertically in a 5 × 10 arrangement. Holes in top and middle deck are 16 mm diam., and holes in lower deck are tapered from 16 to 6 mm diam. Lower deck prevents pipette tips from touching table beneath.
법적 정보
Scienceware is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)