주요 문서
SYNM250005
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 5, Hose Connections: Front-left-right, Rear-left-right
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
40161501
NACRES:
NB.51
특징
number of ports 5
포장
pkg of 1 ea
제조업체/상표
Synthware M250005
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
법적 정보
Synthware is a trademark of Kemtech America Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)