주요 문서
STW6
COware gas reactor
reaction scale of 5.0-20.0 mmol (total volume 400 mL)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103817
NACRES:
NB.51
포장
pack of 1 ea
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Two-chamber glass apparatus are ideal for reaction scales of:
  • 0.1-1 mmol (total volume 20 mL)
  • 1.0-5.0 mmol (total volume 100 mL)
  • 5.0-20.0 mmol (total volume 400 mL)
Each unit includes two screw caps, two stabilizing PTFE discs (recyclable), ten single use silicone/PTFE septa (in combination with the stabilizing discs each septum allows for a maximum of 2 piercings using 21G/Ø0.80 mm syringes or smaller).
Order replacement PTFE/silicone septa STW3.
Learn more about CO releasing molecules from Prof. Skrydstrup
COware gas reactor is a part of the COware Platform
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서
Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.