주요 문서
DWK62113D-2
Kimble® 33 Expansion Tubular Borosilicate Glass Serum Vial Without Closure
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122001
NACRES:
NB.17
material
Borosilicate glass/PTFE tube
tubulated bottom
특징
closure type: no
포장
pkg of 1440 ea
제조업체/상표
Kimble® 62113D-2
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Autoclavable serum vials are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, culture media, chromatography samples, and more.
애플리케이션
Storage and transport of samples, lyophilization, contain harsh reagents or liquids.
특징 및 장점
  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Aluminum seals, stoppers and septa are available separately
  • Converted from Type I borosilicate glass tubing, vials conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)