주요 문서
BR782713
BRAND® 2in1 cell culture inserts
new
24-well, PET, 1µm pore, sterile, multipack (4x12EA), BIO-CERT® quality
동의어(들):
2-in-1, 24-well cell culture insert, 782713, BRAND insert, cell culture membrane, insert, permeable support, transwell
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
material
PET membrane
무균
sterile
포장
pack of 48 ea (4x12 multipack)
제조업체/상표
BRAND 782713
24
공극 크기
1 μm pore size
결합 유형
cell culture treated
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The BRAND® 2in1 BIO-CERT® cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more. Thanks to both supporting feet and hooks, you can position the BRAND® 2in1 BIO-CERT® cell culture insert in a standing or hanging position. Due to its protein-like structure, the cellGrade plus surface provides optimal conditions for growing primary keratinocytes and other cell types. With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row so that the remaining inserts stay sterile.
특징 및 장점
  • Can be used in hanging or standard positions
  • Compatible with all common multiwell plates
  • Suitable for 2D and 3D cell culture
  • PET membrane with cellGrade plus surface
  • Sterility Assurance Level 10-6
  • BIO-CERT® cell culture quality: Free of DNA, RNase, DNase, Pyrogens, and non-cytotoxic
법적 정보
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)