주요 문서
BMSV1002
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Handheld vacuum controller with single channel adapters
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.79
제조업체/상표
Benchmark Scientific V1002
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Accessories designed for use with Aspire laboratory aspirator.
Handheld vacuum controller with single channel adapters.
특징 및 장점
  • The handheld controller can hold 200-300 ul disposable pipette tips
  • The handheld controller and adapters are composed of polyoxymethylene (POM)
  • Autoclave-friendly ergonomic designed for prolonged handheld usage
  • The controller is compatible with serological pipettes and Pasteur pipettes
법적 정보
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)