주요 문서
BMSBSH5002EUK
Sigma-Aldrich
myBlock ll digital dual chamber dry bath without blocks
AC/DC input 230 V AC, UK plug
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41102404
NACRES:
NB.76
제조업체/상표
Benchmark Scientific BSH5002-E-UK
파라미터
5-105 °C temp. range
용량
, holds 2 blocks
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Benchmark′s myBlock series is a "next generation" product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls, timed or continuous operation and a removable hinged lid. In addition, an optional external temperature feedback probe can be positioned directly inside a sample tube, providing real-time control and monitoring of actual sample temperature.
  • Digital temperature control (set and walk away)
  • Hinged lid (improves uniformity & conserves energy)
  • External (optional) temperature probe (measures actual sample temperature)
  • Available with Quick-Flip universal blocks (supports all common tube sizes, PCR strips and PCR plates)
관련 제품
제품 번호
설명
가격
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)