주요 문서
BAH420580001
SP Bel-Art® Dry-Keeper Horizontal Desiccator Cabinet
PVC, volume 2 ft3
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41123002
NACRES:
NB.43
material
PVC
포장
pkg of 1 ea
제조업체/상표
Bel-Art H42058-0001
용적
2 ft3
일반 설명
Clear and durable PVC desiccator cabinet has sleek black ABS accents, horizontal profile, and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
  • Spacious 2 cu. ft. interior volume
  • Two adjustable, removable, and perforated shelves facilitate full circulation of air; shelf rails can be repositioned on any one of 8 steps set at 28mm intervals; additional shelves and rails sold separately
  • Large, in-door digital hygrometer for easy visualization of rH%
  • Gasketed door opens down and closes with two latches for a tight seal; interior opening measures 45 x 23cm (17¾ x 9")
  • Includes desiccant tray
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)